Methyl-5-brom-6-hydroxynicotinat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 5-bromo-6-hydroxynicotinate
PY-7816-1G
PY-7816-5G
COBBPY-7816-1GEA
116
EUR
COBBPY-7816-1G
COBBPY-7816-5G
Methyl-5-brom-6-hydroxynicotinat
Methyl-5-brom-6-hydroxynicotinat
Formel:
C₇H₆BrNO₃ Molecular Weight: 232,03 g/mol |
MDL:
MFCD08235247 CAS-Nummer: 381247-99-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...