Methyl-5-fluornicotinat
Lieferant: COMBI-BLOCKS
Synonyme:
5-Fluoro-nicotinic acid methyl ester, Methyl 5-fluoro-3-pyridinecarboxylate
PY-7852-1G
PY-7852-5G
COBBPY-7852-1GEA
63.2
EUR
COBBPY-7852-1G
COBBPY-7852-5G
Methyl-5-fluornicotinat
Methyl-5-fluornicotinat
Formel:
C₇H₆FNO₂ Molecular Weight: 155,13 g/mol |
MDL:
MFCD01692049 CAS-Nummer: 455-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...