5-Nitropicolinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Nitro-2-pyridinecarboxylic acid, 5-Nitropyridine-2-carboxylic acid, 5-Nitropicolinic acid

PY-7964-1G PY-7964-5G
COBBPY-7964-1GEA 38.2 EUR
COBBPY-7964-1G COBBPY-7964-5G
5-Nitropicolinsäure
5-Nitropicolinsäure
Formel: C₆H₄N₂O₄
Molecular Weight: 168,11 g/mol
MDL: MFCD04114182
CAS-Nummer: 30651-24-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR