Methyl-2-fluornicotinat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 2-fluoronicotinate, 2-Fluoronicotinic acid methyl ester
SS-3007-5G
SS-3007-1G
COBBSS-3007-5GEA
102
EUR
COBBSS-3007-5G
COBBSS-3007-1G
Methyl-2-fluornicotinat
Methyl-2-fluornicotinat
Formel:
C₇H₆FNO₂ Molecular Weight: 155,13 g/mol |
MDL:
MFCD03095072 CAS-Nummer: 446-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...