2-Bromethylethylether
Lieferant: FLUOROCHEM
001004-100G
001004-25G
FLUO001004-100GEA
338
EUR
FLUO001004-100G
FLUO001004-25G
2-Bromethylethylether
2-Bromethylethylether
Formel:
C₄H₉BrO Molecular Weight: 153,02 g/mol |
MDL:
MFCD00000237 CAS-Nummer: 592-55-2 EINECS: 209-763-6 UN: 2340 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...