Cysteaminiumchlorid

Lieferant: FLUOROCHEM

Synonyme: 2-MEA

003420-25G 003420-250G 003420-50G
FLUO003420-25GEA 42.9 EUR
FLUO003420-25G FLUO003420-250G FLUO003420-50G
Cysteaminiumchlorid
Cysteaminiumchlorid
Formel: C₂H₇NS·HCl
Molecular Weight: 113.61 g/mol
MDL: MFCD00012904
CAS-Nummer: 156-57-0
EINECS: 205-858-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR