3-Fluor-2-(trifluormethyl)benzamid

Lieferant: FLUOROCHEM

005814-1G 005814-2.5G
FLUO005814-1GEA 296 EUR
FLUO005814-1G FLUO005814-2.5G
3-Fluor-2-(trifluormethyl)benzamid
3-Fluor-2-(trifluormethyl)benzamid
Formel: C₈H₅F₄NO
Molecular Weight: 207.13 g/mol
MDL: MFCD01631492
CAS-Nummer: 287398-80-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR