4'-Fluor-2'-nitroacetanilid
Lieferant: FLUOROCHEM
Synonyme:
N-(4-Fluoro-2-nitrophenyl)acetamide
006291-25G
006291-100G
FLUO006291-25GEA
137
EUR
FLUO006291-25G
FLUO006291-100G
4'-Fluor-2'-nitroacetanilid
4'-Fluor-2'-nitroacetanilid
Formel:
C₈H₇FN₂O₃ |
MDL:
MFCD00031517 CAS-Nummer: 448-39-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...