2-(2-Fluorphenoxy)-3-nitropyridin

Lieferant: FLUOROCHEM

006320-1G 006320-5G
FLUO006320-1GEA 67.1 EUR
FLUO006320-1G FLUO006320-5G
2-(2-Fluorphenoxy)-3-nitropyridin
2-(2-Fluorophenoxy)-3-nitropyridine
Formel: C₁₁H₇FN₂O₃
Molecular Weight: 234.19 g/mol
MDL: MFCD00067805
CAS-Nummer: 175135-65-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR