2-Fluor-3-(trifluormethyl)benzamid
Lieferant: FLUOROCHEM
CFF1153.
006387-1G
006387-5G
006387-10G
CHMPCFF1153.EA
0
EUR
CHMPCFF1153.
FLUO006387-1G
FLUO006387-5G
FLUO006387-10G
2-Fluor-3-(trifluormethyl)benzamid
2-Fluor-3-(trifluormethyl)benzamid
Formel:
C₈H₅F₄NO Molecular Weight: 207,13 g/mol |
MDL:
MFCD00061146 CAS-Nummer: 207853-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...