2-Fluor-4-(trifluormethyl)benzamid

Lieferant: FLUOROCHEM

006388-1G 006388-5G 006388-10G
FLUO006388-1GEA 39.9 EUR
FLUO006388-1G FLUO006388-5G FLUO006388-10G
2-Fluor-4-(trifluormethyl)benzamid
2-Fluor-4-(trifluormethyl)benzamid
Formel: C₈H₅F₄NO
Molecular Weight: 207.13 g/mol
MDL: MFCD00061147
CAS-Nummer: 207853-64-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR