2-Fluor-6-(trifluormethyl)benzamid

Lieferant: FLUOROCHEM

006390-1G 006390-5G 006390-10G
FLUO006390-1GEA 42.9 EUR
FLUO006390-1G FLUO006390-5G FLUO006390-10G
2-Fluor-6-(trifluormethyl)benzamid
2-Fluor-6-(trifluormethyl)benzamid
Formel: C₈H₅F₄NO
Molecular Weight: 207.13 g/mol
MDL: MFCD00083555
CAS-Nummer: 144851-59-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR