3-Fluor-5-(trifluormethyl)benzamid

Lieferant: FLUOROCHEM

006391-1G 006391-5G 006391-10G
FLUO006391-1GEA 34.1 EUR
FLUO006391-1G FLUO006391-5G FLUO006391-10G
3-Fluor-5-(trifluormethyl)benzamid
3-Fluor-5-(trifluormethyl)benzamid
Formel: C₈H₅F₄NO
Molecular Weight: 207.13 g/mol
MDL: MFCD00061149
CAS-Nummer: 207986-20-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR