4-Fluor-2-(trifluormethyl)benzamid

Lieferant: FLUOROCHEM

006392-1G 006392-5G 006392-10G
FLUO006392-1GEA 34.1 EUR
FLUO006392-1G FLUO006392-5G FLUO006392-10G
4-Fluor-2-(trifluormethyl)benzamid
4-Fluor-2-(trifluormethyl)benzamid
Formel: C₈H₅F₄NO
MDL: MFCD00061150
CAS-Nummer: 207919-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR