2-Acetamido-5-fluorbenzoesäure

Lieferant: FLUOROCHEM

CFF527.2.5 009001-1G 009001-5G
CHMPCFF527.2.5EA 508 EUR
CHMPCFF527.2.5 FLUO009001-1G FLUO009001-5G
2-Acetamido-5-fluorbenzoesäure
2-Acetamido-5-fluorbenzoesäure
Formel: C₉H₈FNO₃
MDL: MFCD00973900
CAS-Nummer: 49579-56-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR