Methyl-5-hydroxynicotinat

Lieferant: FLUOROCHEM

Synonyme: Methyl 5-hydroxynicotinate, Methyl 5-hydroxy-3-pyridinecarboxylate

009282-1G 009282-5G
FLUO009282-1GEA 48.6 EUR
FLUO009282-1G FLUO009282-5G
Methyl-5-hydroxynicotinat
Methyl-5-hydroxynicotinat
Formel: C₇H₇NO₃
Molecular Weight: 153,14 g/mol
MDL: MFCD00191521
CAS-Nummer: 30766-22-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR