3-Chlorpicolinsäure

Lieferant: CHEMPUR

Synonyme: 3-Chloropyridine-2-carboxylic acid

FL10712.POR 010712-1G
CHMPFL10712.POREA 0 EUR
CHMPFL10712.POR FLUO010712-1G
3-Chlorpicolinsäure
3-Chlorpicolinsäure
Formel: C₆H₄ClNO₂
Molecular Weight: 157,56 g/mol
MDL: MFCD01646325
CAS-Nummer: 57266-69-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR