3-Fluorisonicotinsäure

Lieferant: FLUOROCHEM

Synonyme: 3-Fluoropyridine-4-carboxylic acid

010728-1G 010728-5G
FLUO010728-1GEA 31.1 EUR
FLUO010728-1G FLUO010728-5G
3-Fluorisonicotinsäure
3-Fluorisonicotinsäure
Formel: C₆H₄FNO₂
Molecular Weight: 141,1 g/mol
MDL: MFCD03789251
CAS-Nummer: 393-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR