5-Chlorisatosäureanhydrid
Lieferant: FLUOROCHEM
Synonyme:
6-Chloro-1H-benzo[d][1,3]oxazine-2,4-dione
011637-10G
011637-100G
011637-25G
FLUO011637-10GEA
0
EUR
FLUO011637-10G
FLUO011637-100G
FLUO011637-25G
5-Chlorisatosäureanhydrid
5-Chlorisatosäureanhydrid
Formel:
C₈H₄ClNO₃ Molecular Weight: 197,58 g/mol |
MDL:
MFCD00006701 CAS-Nummer: 4743-17-3 EINECS: 225-255-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...