Diphenyliodoniumbromid

Lieferant: FLUOROCHEM

013149-5G 013149-25G
FLUO013149-5GEA 411 EUR
FLUO013149-5G FLUO013149-25G
Diphenyliodoniumbromid
Diphenyliodonium bromide
Formel: C₁₂H₁₀BrI
MDL: MFCD00031703
CAS-Nummer: 1483-73-4
EINECS: 216-050-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR