2,3,5-Trifluorisonicotinsäure
Lieferant: FLUOROCHEM
Synonyme:
2,3,5-Trifluoropyridine-4-carboxylic acid
013179-1G
013179-5G
FLUO013179-1GEA
0
EUR
FLUO013179-1G
FLUO013179-5G
2,3,5-Trifluorisonicotinsäure
2,3,5-Trifluorisonicotinsäure
Formel:
C₆H₂F₃NO₂ Molecular Weight: 177,08 g/mol |
MDL:
MFCD03840205 CAS-Nummer: 675602-91-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...