2,3,6-Trifluorisonicotinsäure
Lieferant: FLUOROCHEM
Synonyme:
2,3,6-Trifluoropyridine-4-carboxylic acid
013180-1G
013180-5G
FLUO013180-1GEA
0
EUR
FLUO013180-1G
FLUO013180-5G
2,3,6-Trifluorisonicotinsäure
2,3,6-Trifluorisonicotinsäure
Formel:
C₆H₂F₃NO₂ Molecular Weight: 177.08 g/mol |
MDL:
MFCD03840204 CAS-Nummer: 675602-92-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...