Ethyl-6-chlornicotinat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 6-chloronicotinate

013880-5G 013880-25G
FLUO013880-5GEA 0 EUR
FLUO013880-5G FLUO013880-25G
Ethyl-6-chlornicotinat
Ethyl-6-chlornicotinat
Formel: C₈H₈ClNO₂
MDL: MFCD00082739
CAS-Nummer: 49608-01-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR