2,6-Dichlorisonicotinsäure

Lieferant: FLUOROCHEM

Synonyme: 2,6-Dichloropyridine-4-carboxylic acid

017205-2.5G 017205-25G 017205-5G
FLUO017205-2.5GEA 0 EUR
FLUO017205-2.5G FLUO017205-25G FLUO017205-5G
2,6-Dichlorisonicotinsäure
2,6-Dichlorisonicotinsäure
Formel: C₆H₃Cl₂NO₂
Molecular Weight: 192 g/mol
MDL: MFCD00234147
CAS-Nummer: 5398-44-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR