5-Brom-2-hydroxy-3-nitropyridin
Lieferant: FLUOROCHEM
017235-5G
017235-1G
FLUO017235-5GEA
39.9
EUR
FLUO017235-5G
FLUO017235-1G
5-Brom-2-hydroxy-3-nitropyridin
5-Brom-2-hydroxy-3-nitropyridin
Formel:
C₅H₃BrN₂O₃ |
MDL:
MFCD00023473 CAS-Nummer: 15862-34-7 EINECS: 239-989-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...