Methyl-5-bromnicotinat

Lieferant: FLUOROCHEM

CFF357.5 CFF357.25 017390-5G 017390-25G
CHMPCFF357.5EA 321 EUR
CHMPCFF357.5 CHMPCFF357.25 FLUO017390-5G FLUO017390-25G
Methyl-5-bromnicotinat
Methyl-5-bromnicotinat
Formel: C₇H₆BrNO₂
MDL: MFCD00173668
CAS-Nummer: 29681-44-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR