Acetamidin Hydrochlorid

Lieferant: FLUOROCHEM

021800-500G 021800-1KG
FLUO021800-500GEA 251 EUR
FLUO021800-500G FLUO021800-1KG
Acetamidin Hydrochlorid
Acetamidin Hydrochlorid
Formel: C₂H₇ClN₂
MDL: MFCD00013016
CAS-Nummer: 124-42-5
EINECS: 204-700-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR