5-Brom-2-pyridincarbonitril

Lieferant: FLUOROCHEM

Synonyme: 5-Bromopicolinonitrile, 5-Bromo-2-cyanopyridine

045207-25G 045207-5G
FLUO045207-25GEA 119 EUR
FLUO045207-25G FLUO045207-5G
5-Brom-2-pyridincarbonitril
5-Brom-2-pyridincarbonitril
Formel: C₆H₃BrN₂
Molecular Weight: 183,01 g/mol
MDL: MFCD00234144
CAS-Nummer: 97483-77-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR