4-(4-Nitrobenzyloxy)benzaldehyd

Lieferant: MENAI

IP60.50 IP60.100
MENAIP60.50EA 0 EUR
MENAIP60.50 MENAIP60.100
4-(4-Nitrobenzyloxy)benzaldehyd
4-(4-Nitrobenzyloxy)benzaldehyd
Formel: C₁₄H₁₁NO₄
Molecular Weight: 257.25 g/mol
MDL: MFCD00017041
CAS-Nummer: 67565-48-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR