Triethanolamin Hydrochlorid, Millipore®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

1.08357.0100 1.08357.1000
1.08357.0100EA 48.4 EUR
1.08357.0100 1.08357.1000
Triethanolamin Hydrochlorid, Millipore®
Triethanolamin Hydrochlorid
Formel: C₆H₁₆ClNO₃
Molecular Weight: 185,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012596
CAS-Nummer: 637-39-8
EINECS: 211-284-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR