Aminoguanidiniumhydrogencarbonat, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Aminoguanidine hydrogen carbonate, Aminoguanidine bicarbonate, 1-Aminoguanidinium hydrogen carbonate, Aminoguanidine hydrogencarbonate, Guanylhydrazine hydrogencarbonate

M-Clarity™ Qualitätsstufe = MQ200

8.01417.0250 8.01417.9050 8.01417.9025
8.01417.0250EA 39.2 EUR
8.01417.0250 8.01417.9050 8.01417.9025
Aminoguanidiniumhydrogencarbonat, Sigma-Aldrich®
Aminoguanidinium hydrogen carbonate
Formel: C₂H₈N₄O₃
Molecular Weight: 136.11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012949
CAS-Nummer: 2582-30-1
EINECS: 219-956-7
UN: 3077
ADR: 9 III

Bestellen

Spezifikation Testergebnisse

Solubility  - 5 g/l (20 °C)
Melting Point 125 °C decomposes
Molar Mass 136.11 g/mol
Bulk Density 700 kg/m3
Density 1.56 g/cm3 (20 °C)
pH 8.9 (5 g/l, H2O, 20 °C)
Ignition Point 245 °C
Synonyms Aminoguanidine bicarbonate
Hill Formula C2H8N4O3
Chemical Formula CH6N4 * H2CO3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR