Hydantoin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Imidazolidine-2,4-dione
M-Clarity™ Qualitätsstufe = MQ200
8.04511.0100
8.04511.0500
8.04511.0100EA
36.4
EUR
8.04511.0100
8.04511.0500
Hydantoin, Sigma-Aldrich®
Hydantoin
Formel:
C₃H₄N₂O₂ Molecular Weight: 100,08 g/mol |
MDL:
MFCD00005259 CAS-Nummer: 461-72-3 EINECS: 207-313-3 |
Spezifikation Testergebnisse
Solubility | (20 °C) slightly soluble |
Melting Point | 219 - 222 °C |
Molar Mass | 100.08 g/mol |
Synonyms | 2,4-(3H,5H)-Imidazoledione |
Hill Formula | C3H4N2O2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...