Ethylenglycolmonoisopropylether, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
2-Isopropoxyethanol
M-Clarity™ Qualitätsstufe = MQ200
8.07480.1000
8.07480.9020
8.07480.1000EA
34.3
EUR
8.07480.1000
8.07480.9020
Ethylenglycolmonoisopropylether, Sigma-Aldrich®
Ethylenglycolmonoisopropylether
Formel:
C₅H₁₂O₂ Molecular Weight: 104.15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002866 CAS-Nummer: 109-59-1 EINECS: 203-685-6 UN: 3271 ADR: 3 III |
Spezifikation Testergebnisse
Solubility | (20 °C) soluble |
Molar Mass | 104.15 g/mol |
Boiling Point | 143 °C (1013 hPa) |
Vapor Pressure | 3.5 hPa (20 °C) |
Flash Point | 45 °C |
Refractive Index | 1.41 (20 °C, 589 nm) |
Explosion Limit | 1.6 - 13.0 %(V) |
Density | 0.90 g/cm3 (20 °C) |
pH | 7 (H2O, 20 °C) |
Ignition Point | 240 °C |
Evaporation number | 52 |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...