Ethylenglycolmonoisopropylether, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Isopropoxyethanol

M-Clarity™ Qualitätsstufe = MQ200

107891-1L 107891-25ML 107891-2L
SIAL107891-1LEA 65.6 EUR
SIAL107891-1L SIAL107891-25ML SIAL107891-2L
Ethylenglycolmonoisopropylether, Sigma-Aldrich®
Ethylenglycolmonoisopropylether
Formel: C₅H₁₂O₂
Molecular Weight: 104.15 g/mol
MDL: MFCD00002866
CAS-Nummer: 109-59-1
EINECS: 203-685-6
UN: 1992
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR