Aminoguanidiniumhydrogencarbonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Aminoguanidine hydrogen carbonate, Aminoguanidine bicarbonate, 1-Aminoguanidinium hydrogen carbonate, Aminoguanidine hydrogencarbonate, Guanylhydrazine hydrogencarbonate

M-Clarity™ Qualitätsstufe = MQ100

109266-100G 109266-500G
SIAL109266-100GEA 25.1 EUR
SIAL109266-100G SIAL109266-500G
Aminoguanidiniumhydrogencarbonat, Sigma-Aldrich®
Aminoguanidinium hydrogen carbonate
Formel: C₂H₈N₄O₃
Molecular Weight: 136.11 g/mol
MDL: MFCD00012949
CAS-Nummer: 2582-30-1
EINECS: 219-956-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR