Aminoacetaldehyddimethylacetal, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2-Dimethoxyethanamine

M-Clarity™ Qualitätsstufe = MQ200

121967-100ML 121967-25ML 121967-500ML
SIAL121967-100MLEA 143 EUR
SIAL121967-100ML SIAL121967-25ML SIAL121967-500ML
Aminoacetaldehyddimethylacetal, Sigma-Aldrich®
Aminoacetaldehyddimethylacetal
Formel: C₄H₁₁NO₂
Molecular Weight: 105,14 g/mol
MDL: MFCD00008135
CAS-Nummer: 22483-09-6
EINECS: 245-026-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR