Allylcyanid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

122793-100G 122793-5G
SIAL122793-100GEA 339 EUR
SIAL122793-100G SIAL122793-5G
Allylcyanid, Sigma-Aldrich®
Allylcyanid
Formel: C₄H₅N
Molecular Weight: 67,09 g/mol
MDL: MFCD00001962
CAS-Nummer: 109-75-1
EINECS: 203-701-1
UN: 1992
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR