cis-1,2,3,6-Tetrahydrophthalsäureanhydrid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: cis-4-Cyclohexene-1,2-dicarboxylic anhydride

M-Clarity™ Qualitätsstufe = MQ100

136891-100G 136891-1KG 136891-2.5KG
SIAL136891-100GEA 59.6 EUR
SIAL136891-100G SIAL136891-1KG SIAL136891-2.5KG
cis-1,2,3,6-Tetrahydrophthalsäureanhydrid, Sigma-Aldrich®
cis-1,2,3,6-Tetrahydrophthalsäureanhydrid
Formel: C₈H₈O₃
Molecular Weight: 152,15 g/mol
MDL: MFCD00005916
CAS-Nummer: 935-79-5
EINECS: 213-308-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR