1,8-Diazabicyclo[5.4.0]-7-undecen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,3,4,6,7,8,9,10-Octahydropyrimido[1,2-a]azepine, DBU

M-Clarity™ Qualitätsstufe = MQ200

139009-100G 139009-25G 139009-500G
SIAL139009-100GEA 85.8 EUR
SIAL139009-100G SIAL139009-25G SIAL139009-500G
1,8-Diazabicyclo[5.4.0]-7-undecen, Sigma-Aldrich®
1,8-Diazabicyclo[5.4.0]-7-undecen
Formel: C₉H₁₆N₂
Molecular Weight: 152,24 g/mol
MDL: MFCD00006930
CAS-Nummer: 6674-22-2
EINECS: 229-713-7
UN: 3267
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR