1-(1-Cyclopenten-1-yl)pyrrolidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Pyrrolidino-1-cyclopentene

149446-10G 149446-50G
SIAL149446-10GEA 56.7 EUR
SIAL149446-10G SIAL149446-50G
1-(1-Cyclopenten-1-yl)pyrrolidin, Sigma-Aldrich®
1-(1-Cyclopenten-1-yl)pyrrolidin
Formel: C₉H₁₅N
Molecular Weight: 137,22 g/mol
MDL: MFCD00003162
CAS-Nummer: 7148-07-4
EINECS: 230-463-6
UN: 1993
ADR: 3,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR