Diethyl-3-oxoglutarat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diethyl 1,3-acetonedicarboxylate, Diethyl acetone-1,3-dicarboxylate

M-Clarity™ Qualitätsstufe = MQ100

165123-250G 165123-50G
SIAL165123-250GEA 196 EUR
SIAL165123-250G SIAL165123-50G
Diethyl-3-oxoglutarat, Sigma-Aldrich®
Diethyl-3-oxoglutarat
Formel: C₉H₁₄O₅
Molecular Weight: 202,21 g/mol
MDL: MFCD00009200
CAS-Nummer: 105-50-0
EINECS: 203-302-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR