Bis(trimethylsilyl)acetylen, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
187437-10G
187437-50G
SIAL187437-10GEA
70.4
EUR
SIAL187437-10G
SIAL187437-50G
Bis(trimethylsilyl)acetylen, Sigma-Aldrich®
Bis(trimethylsilyl)acetylen
Formel:
C₈H₁₈Si₂ Molecular Weight: 170.4 g/mol |
MDL:
MFCD00008276 CAS-Nummer: 14630-40-1 EINECS: 238-671-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...