L(+)-α-Phenylglycin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: H-Phg-OH, L-(+)-2-Phenylglycine

M-Clarity™ Qualitätsstufe = MQ100

237647-100G 237647-25G
SIAL237647-100GEA 79.8 EUR
SIAL237647-100G SIAL237647-25G
L(+)-α-Phenylglycin, Sigma-Aldrich®
L(+)-α-Phenylglycin
Formel: C₈H₉NO₂
Molecular Weight: 151,16 g/mol
MDL: MFCD00064403
CAS-Nummer: 2935-35-5
EINECS: 220-909-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR