Cystaminiumdichlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Cystaminium dichloride, 2,2′-Diaminodiethyl disulphide dihydrochloride, 2,2'-Disulphanediyldiethanamine dihydrochloride, Bis(2-aminoethyl) disulphide dihydrochloride, Decarboxycystine dihydrochloride, Cystamine (dihydrochloride), {2-[(2-Aminoethyl)disulphanyl]ethyl}amine dihydrochloride, 2,2′-Dithiobis(ethylamine) dihydrochloride
M-Clarity™ Qualitätsstufe = MQ200
30050-100G-F
30050-25G-F
SIAL30050-100G-FEA
169
EUR
SIAL30050-100G-F
SIAL30050-25G-F
Cystaminiumdichlorid, Sigma-Aldrich®
Cystaminiumdichlorid
Formel:
C₄H₁₂N₂S₂·2HCl Molecular Weight: 225.20 g/mol |
MDL:
MFCD00012905 CAS-Nummer: 56-17-7 EINECS: 200-260-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...