Gallium(III)chlorid, ultra trocken ≥99,999% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Gallium trichloride
Unter Argon in Ampullen verpackt
43879.06
43879.14
43879.22
43879.06EA
88.9
EUR
43879.06
43879.14
43879.22
Gallium(III)chlorid, ultra trocken ≥99,999% (Metall-Basis)
Gallium(III)chlorid
Purity: 99.999%
CAS No.: 13450-90-3
Molecular Formula: GaCl3
Formula Weight: 176.03
Physical Form: Crystalline, Ampouled under argon
Density: 2.47
Appearance: White
Melting Point: 78°
Boiling Point: 201°
Merck Reference: 14,4348
UN No.: 3260
MDL No.: MFCD00011018
Formel:
Cl₃Ga Molecular Weight: 176,08 g/mol Siedepunkt: 201 °C Schmelzpunkt: 78 °C Dichte: 2,47 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011018 CAS-Nummer: 13450-90-3 EINECS: 236-610-0 UN: 3260 ADR: 8,II Merck Index: 14,4348 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...