N-Methylpropylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Methyl-1-propylamine

308137-1G 308137-5G
SIAL308137-1GEA 24.9 EUR
SIAL308137-1G SIAL308137-5G
N-Methylpropylamin, Sigma-Aldrich®
N-Methylpropylamin
Formel: C₄H₁₁N
Molecular Weight: 73.14 g/mol
MDL: MFCD00009361
CAS-Nummer: 627-35-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR