1,8-Diazabicyclo[5.4.0]-7-undecen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,3,4,6,7,8,9,10-Octahydropyrimido[1,2-a]azepine, DBU

M-Clarity™ Qualitätsstufe = MQ200

33482-1L-F 33482-250ML-F 33482-50ML-F
SIAL33482-1L-FEA 670 EUR
SIAL33482-1L-F SIAL33482-250ML-F SIAL33482-50ML-F
1,8-Diazabicyclo[5.4.0]-7-undecen, Sigma-Aldrich®
1,8-Diazabicyclo[5.4.0]-7-undecen
Formel: C₉H₁₆N₂
Molecular Weight: 152,24 g/mol
MDL: MFCD00006930
CAS-Nummer: 6674-22-2
EINECS: 229-713-7
UN: 3267
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR