Pentaethylenglycol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3,6,9,12-Tetraoxatetradecane-1,14-diol
M-Clarity™ Qualitätsstufe = MQ200
335754-25G
335754-5G
SIAL335754-25GEA
656
EUR
SIAL335754-25G
SIAL335754-5G
Pentaethylenglycol, Sigma-Aldrich®
Pentaethylenglycol
Formel:
C₁₀H₂₂O₆ Molecular Weight: 238,28 g/mol |
MDL:
MFCD00002878 CAS-Nummer: 4792-15-8 EINECS: 225-341-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...