1-Phenyl-2-(trimethylsilyl)acetylen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Trimethyl(phenylethynyl)silane

M-Clarity™ Qualitätsstufe = MQ100

338923-10G 338923-50G
SIAL338923-10GEA 157 EUR
SIAL338923-10G SIAL338923-50G
1-Phenyl-2-(trimethylsilyl)acetylen, Sigma-Aldrich®
1-Phenyl-2-(trimethylsilyl)acetylen
Formel: C₁₁H₁₄Si
Molecular Weight: 174,32 g/mol
MDL: MFCD00054860
CAS-Nummer: 2170-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR