(Triisopropylsilyl)acetylen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethynyltriisopropylsilane

M-Clarity™ Qualitätsstufe = MQ100

360031-25G 360031-5G
SIAL360031-25GEA 317 EUR
SIAL360031-25G SIAL360031-5G
(Triisopropylsilyl)acetylen, Sigma-Aldrich®
(Triisopropylsilyl)acetylen
Formel: C₁₁H₂₂Si
Molecular Weight: 182,38 g/mol
MDL: MFCD00075452
CAS-Nummer: 89343-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR